ENAH31552319 Enamine

(4-ethyl-2-fluorophenyl)methanamine hydrochloride

  • MDL number MFCD31794607
  • Cas Number : 2287281-37-0
  • Molecular Weight : 189.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744170
Availability details for
Quantity
  • Availability
  • Add to Cart