ENAH31552322 Enamine

3-(tert-butoxy)-2-(prop-2-en-1-yl)cyclobutan-1-amine

  • MDL number MFCD31794440
  • Cas Number : 2287283-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745327
Availability details for
Quantity
  • Availability
  • Add to Cart