ENAH31552323 Enamine

3-[(benzyloxy)methyl]spiro[3.3]heptan-1-one

  • MDL number MFCD31794441
  • Cas Number : 2287330-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745342
Availability details for
Quantity
  • Availability
  • Add to Cart