ENAH31552325 Enamine

2-[2-(2-hydroxyethyl)-1-benzothiophen-3-yl]ethan-1-ol

  • MDL number MFCD31794443
  • Cas Number : 2287298-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745364
Availability details for
Quantity
  • Availability
  • Add to Cart