ENAH31552329 Enamine

(2R)-1-(6-methoxynaphthalen-2-yl)propan-2-amine hydrochloride

  • MDL number MFCD31794662
  • Cas Number : 2287247-04-3
  • Molecular Weight : 251.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6745561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745561
Availability details for
Quantity
  • Availability
  • Add to Cart