ENAH3155232D Enamine

3-methoxy-2,2,4,4-tetramethylpyrrolidine

  • MDL number MFCD31794795
  • Cas Number : 2287339-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745672
Availability details for
Quantity
  • Availability
  • Add to Cart