ENAH31552334 Enamine

ethyl 2-cyano-1-(propan-2-yl)cyclopropane-1-carboxylate

  • MDL number MFCD31794699
  • Cas Number : 2287273-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745742
Availability details for
Quantity
  • Availability
  • Add to Cart