ENAH31552338 Enamine

1-(bromomethyl)-4-(difluoromethylidene)cyclohexane

  • MDL number MFCD31794702
  • Cas Number : 2287310-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745794
Availability details for
Quantity
  • Availability
  • Add to Cart