ENAH3155233A Enamine

2-{1-[(tert-butoxy)carbonyl]-2,5-dihydro-1H-pyrrol-3-yl}-2-methylpropanoic acid

  • MDL number MFCD31794706
  • Cas Number : 2287344-34-5
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745860
Availability details for
Quantity
  • Availability
  • Add to Cart