ENAH31552346 Enamine

1-methoxybicyclo[3.3.1]nonan-3-amine hydrochloride

  • MDL number MFCD31794553
  • Cas Number : 2287343-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746175
Availability details for
Quantity
  • Availability
  • Add to Cart