ENAH3155234B Enamine

methyl (2S,4R)-4-ethoxypyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD31794719
  • Cas Number : 2287249-69-6
  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746260
Availability details for
Quantity
  • Availability
  • Add to Cart