ENAH3155234C Enamine

{2-aminospiro[3.3]heptan-2-yl}methanol hydrochloride

  • MDL number MFCD31794602
  • Cas Number : 2287301-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746286
Availability details for
Quantity
  • Availability
  • Add to Cart