ENAH3155234F Enamine

5-phenyl-1,2,3-thiadiazole-4-sulfonyl chloride

  • MDL number MFCD31794721
  • Cas Number : 2287322-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746299
Availability details for
Quantity
  • Availability
  • Add to Cart