ENAH31552355 Enamine

2-(iodomethyl)-hexahydrofuro[3,2-b]furan

  • MDL number MFCD31794726
  • Cas Number : 2287286-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -70
Physical Form liquid
Shipping Temperature Dry Ice
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746440

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6746440
pictograms GHS05,GHS07
hstatements H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart