ENAH31552356 Enamine

tert-butyl (3R)-4-(chlorosulfonyl)-3-methylpiperazine-1-carboxylate

  • MDL number MFCD31794644
  • Cas Number : 2287237-09-4
  • Molecular Weight : 298.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746463
Availability details for
Quantity
  • Availability
  • Add to Cart