ENAH31552368 Enamine

tert-butyl N-(3-bromo-5-cyclopropyl-1,2-oxazol-4-yl)carbamate

  • MDL number MFCD31795000
  • Cas Number : 2287341-85-7
  • Molecular Weight : 303.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746699
Availability details for
Quantity
  • Availability
  • Add to Cart