ENAH31552369 Enamine

5-[(pyrrolidin-3-yloxy)methyl]-1H-1,2,3,4-tetrazole hydrochloride

  • MDL number MFCD31795003
  • Cas Number : 2287281-33-6
  • Molecular Weight : 205.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746736
Availability details for
Quantity
  • Availability
  • Add to Cart