ENAH3155236D Enamine

2-{4-amino-1H,4H,5H,6H,7H,8H-cyclohepta[c]pyrazol-1-yl}ethan-1-ol dihydrochloride

  • MDL number MFCD31795016
  • Cas Number : 2287317-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746788
Availability details for
Quantity
  • Availability
  • Add to Cart