ENAH31552373 Enamine

ethyl (1s,3s)-3-fluoro-3-(hydroxymethyl)cyclobutane-1-carboxylate

  • Cas Number : 2287290-10-0
  • Molecular Weight : 176.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746917
Availability details for
Quantity
  • Availability
  • Add to Cart