ENAH3155237B Enamine

tert-butyl (2S)-2-{[4-(aminomethyl)-1H-1,2,3-triazol-1-yl]methyl}pyrrolidine-1-carboxylate hydrochloride

  • MDL number MFCD31794771
  • Cas Number : 2287237-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748205
Availability details for
Quantity
  • Availability
  • Add to Cart