ENAH31552382 Enamine

[5-(2-methoxyphenyl)-1,3-oxazol-2-yl]methanol

  • MDL number MFCD31794446
  • Cas Number : 1894059-09-6
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745395
Availability details for
Quantity
  • Availability
  • Add to Cart