ENAH31552386 Enamine

methyl 8-oxa-1-azaspiro[4.5]decane-3-carboxylate hydrochloride

  • MDL number MFCD31794826
  • Cas Number : 2287318-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743493
Availability details for
Quantity
  • Availability
  • Add to Cart