ENAH31552387 Enamine

tert-butyl (3R,5S)-3,5-dimethoxymorpholine-4-carboxylate

  • MDL number MFCD31794476
  • Cas Number : 2287236-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745442
Availability details for
Quantity
  • Availability
  • Add to Cart