ENAH31552390 Enamine

diethyl (carbamothioylmethyl)phosphonate

  • MDL number MFCD00192020
  • Cas Number : 77679-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745511

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6745511
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart