ENAH31552393 Enamine

5-bromo-3-isocyanato-1-methyl-1H-indole

  • MDL number MFCD31794661
  • Cas Number : 1080028-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745553
Availability details for
Quantity
  • Availability
  • Add to Cart