ENAH31552398 Enamine

2,2-dimethylpiperidin-4-ol hydrochloride

  • MDL number MFCD20529139
  • Cas Number : 1420671-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745571
Availability details for
Quantity
  • Availability
  • Add to Cart