ENAH3155239C Enamine

5-bromo-4-phenyl-1,3-oxazole

  • MDL number MFCD16990233
  • Cas Number : 20756-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745604
Availability details for
Quantity
  • Availability
  • Add to Cart