ENAH315523A0 Enamine

2,2,4,4-tetramethylpyrrolidin-3-ol

  • MDL number MFCD31794562
  • Cas Number : 1784608-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743124
Availability details for
Quantity
  • Availability
  • Add to Cart