ENAH315523A8 Enamine

5-(3-fluoropropyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD24049584
  • Cas Number : 1536385-87-1
  • Molecular Weight : 130.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743553
Availability details for
Quantity
  • Availability
  • Add to Cart