ENAH315523B2 Enamine

octahydro-1H-4lambda6-[1,2,4]thiadiazino[2,3-a]azepine-4,4-dione

  • MDL number MFCD31794574
  • Cas Number : 2287343-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743782
Availability details for
Quantity
  • Availability
  • Add to Cart