ENAH315523B6 Enamine

3-amino-N-ethyl-4-hydroxybenzene-1-sulfonamide hydrochloride

  • MDL number MFCD31794811
  • Cas Number : 2287300-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743291
Availability details for
Quantity
  • Availability
  • Add to Cart