ENAH315523BD Enamine

(1S)-1-[4-(trifluoromethyl)phenyl]ethane-1,2-diol

  • MDL number MFCD31794676
  • Cas Number : 306281-87-8
  • Molecular Weight : 206.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745834
Availability details for
Quantity
  • Availability
  • Add to Cart