ENAH315523BF Enamine

8-oxa-5-azaspiro[3.6]decane hydrochloride

  • MDL number MFCD31794565
  • Cas Number : 2287302-08-1
  • Molecular Weight : 177.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743294
Availability details for
Quantity
  • Availability
  • Add to Cart