ENAH315523C3 Enamine

[1-(2-chlorophenyl)-2-(methylsulfanyl)ethyl](methyl)amine

  • MDL number MFCD16850347
  • Cas Number : 1250976-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743339
Availability details for
Quantity
  • Availability
  • Add to Cart