ENAH315523C5 Enamine

ethyl 2-[(piperidin-3-yl)methyl]-1,3-thiazole-4-carboxylate hydrochloride

  • MDL number MFCD31794679
  • Cas Number : 2287282-13-5
  • Molecular Weight : 290.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745866
Availability details for
Quantity
  • Availability
  • Add to Cart