ENAH315523CA Enamine

2,4-dibromobut-1-ene

  • MDL number MFCD18971835
  • Cas Number : 55030-57-4
  • Molecular Weight : 213.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745914

Safety Information

signalword Danger
pcodes P210,P264,P270,P280,P301+P312,P305+P351+P338,P310,P330,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-6745914
pictograms GHS05,GHS07
hstatements H227,H302,H318
Availability details for
Quantity
  • Availability
  • Add to Cart