ENAH315523D2 Enamine

thieno[3,2-c]pyridine-3-carboxylic acid

  • MDL number MFCD20652922
  • Cas Number : 775530-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743344
Availability details for
Quantity
  • Availability
  • Add to Cart