ENAH315523D7 Enamine

2,2-difluorobut-3-en-1-amine hydrochloride

  • MDL number MFCD31794683
  • Cas Number : 108781-54-0
  • Molecular Weight : 143.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746077
Availability details for
Quantity
  • Availability
  • Add to Cart