ENAH315523E1 Enamine

(3-bromocyclobutyl)methanol

  • MDL number MFCD31794580
  • Cas Number : 1896670-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744059
Availability details for
Quantity
  • Availability
  • Add to Cart