ENAH315523E6 Enamine

[1-(4-chlorophenyl)-3,3-difluorocyclobutyl]methanol

  • MDL number MFCD31794839
  • Cas Number : 1431983-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744064
Availability details for
Quantity
  • Availability
  • Add to Cart