ENAH315523F4 Enamine

2,2,2-trifluoroethyl N-{4H,6H,7H-pyrano[4,3-d][1,3]thiazol-2-yl}carbamate

  • MDL number MFCD28133320
  • Cas Number : 1924283-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746272
Availability details for
Quantity
  • Availability
  • Add to Cart