ENAH315523FB Enamine

1-chlorodifluoromethanesulfonyl-2-fluorobenzene

  • MDL number MFCD27942370
  • Cas Number : 1228781-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746298
Availability details for
Quantity
  • Availability
  • Add to Cart