ENAH315523FD Enamine

[4-(butan-2-yl)phenyl]methanamine hydrochloride

  • MDL number MFCD31794933
  • Cas Number : 59528-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744184
Availability details for
Quantity
  • Availability
  • Add to Cart