ENAH315523FF Enamine

5,7-difluoro-1-methyl-1H-indole-3-carboxylic acid

  • MDL number MFCD31794606
  • Cas Number : 1781178-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746304
Availability details for
Quantity
  • Availability
  • Add to Cart