ENAH31552407 Enamine

2-[7-(benzyloxy)-1H-indol-3-yl]acetic acid

  • MDL number MFCD04972037
  • Cas Number : 99102-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743180
Availability details for
Quantity
  • Availability
  • Add to Cart