ENAH3155240C Enamine

ethyl 3-(2-aminoethyl)-1H-indole-2-carboxylate hydrochloride

  • MDL number MFCD09836804
  • Cas Number : 53590-32-2
  • Molecular Weight : 268.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6746347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746347
Availability details for
Quantity
  • Availability
  • Add to Cart