ENAH31552413 Enamine

tert-butyl N-[3-(aminomethyl)-3-hydroxycyclobutyl]carbamate

  • MDL number MFCD31794643
  • Cas Number : 2287288-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746461
Availability details for
Quantity
  • Availability
  • Add to Cart