ENAH31552415 Enamine

(2-chloro-1H-imidazol-4-yl)methanol

  • MDL number MFCD19227561
  • Cas Number : 1067894-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746466
Availability details for
Quantity
  • Availability
  • Add to Cart