ENAH31552417 Enamine

(2Z)-2-iodobut-2-enedioic acid

  • Cas Number : 5029-68-5
  • Molecular Weight : 241.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-194
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6746479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746479
Availability details for
Quantity
  • Availability
  • Add to Cart