ENAH31552418 Enamine

tert-butyl 4-bromo-3-(pyridin-2-yl)-1H-pyrazole-1-carboxylate

  • MDL number MFCD31794938
  • Cas Number : 2287271-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744469
Availability details for
Quantity
  • Availability
  • Add to Cart